Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
xrn-1
Entrez Gene ID
175059
Gene Name
5'-3' exoribonuclease 1
Gene Aliases
CELE_Y39G8C.1
Chromosome Location
Chr.II: 14068907 - 14089877 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064544.6 NP_496945.3 3 - 4 608 123

View Full Product Details