Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-8
Entrez Gene ID
175108
Gene Name
protein-coding
Gene Aliases
CELE_K10H10.3
Chromosome Location
Chr.II: 14498026 - 14499755 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027079.3 NP_001022250.1 1 - 2 220 66
NM_001381658.1 NP_001368222.1 1 - 2 46 66

View Full Product Details