Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-8
Entrez Gene ID
175108
Gene Name
protein-coding
Gene Aliases
CELE_K10H10.3
Chromosome Location
Chr.II: 14498026 - 14499755 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027079.3 NP_001022250.1 3 - 4 601 109
NM_001306614.3 NP_001293543.1 2 - 3 259 109
NM_001381658.1 NP_001368222.1 3 - 4 427 109
NR_002361.1 3 - 4 479 109

View Full Product Details