Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mog-4
Entrez Gene ID
175117
Gene Name
protein-coding
Gene Aliases
CELE_C04H5.6
Chromosome Location
Chr.II: 14552494 - 14561599 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064626.5 NP_497027.1 3 - 4 1911 134
GenBank mRNA AF286900.1 - 3 - 4 1927 134

View Full Product Details