Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
par-2
Entrez Gene ID
175275
Gene Name
protein-coding
Gene Aliases
CELE_F58B6.3
Chromosome Location
Chr.III: 1081401 - 1095082 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027459.6 NP_001022630.2 6 - 7 1325 64

View Full Product Details