Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-46
Entrez Gene ID
175279
Gene Name
Probable ligand-gated ion channel 46
Gene Aliases
CELE_Y71D11A.5
Chromosome Location
Chr.III: 1125230 - 1136025 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064937.7 NP_497338.2 11 - 12 1463 77

View Full Product Details