Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdh-12
Entrez Gene ID
190583
Gene Name
protein-coding
Gene Aliases
CELE_Y71D11A.1
Chromosome Location
Chr.III: 1152044 - 1181147 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373846.2 NP_001360600.1 7 - 8 2644 72

View Full Product Details