Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eak-3
Entrez Gene ID
190782
Gene Name
Enhancer of AKt-1 null
Gene Aliases
CELE_Y92C3A.3
Chromosome Location
Chr.III: 1175074 - 1176558 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064942.2 NP_497343.1 2 - 3 221 89

View Full Product Details