Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-63
Entrez Gene ID
191040
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y119D3B.7
Chromosome Location
Chr.III: 1243508 - 1245358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064953.2 NP_497354.1 2 - 3 734 93

View Full Product Details