Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-29
Entrez Gene ID
191056
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_ZC47.3
Chromosome Location
Chr.III: 1272474 - 1273700 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027805.3 NP_001022976.1 2 - 3 729 121

View Full Product Details