Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxa-80
Entrez Gene ID
190761
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y82E9BL.14
Chromosome Location
Chr.III: 1313297 - 1315229 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064980.5 NP_497381.2 1 - 2 127 124

View Full Product Details