Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- elc-1
- Entrez Gene ID
-
190775
- Gene Name
- Skp1_POZ domain-containing protein
- Gene Aliases
-
CELE_Y82E9BR.15
- Chromosome Location
-
Chr.III: 1414239 - 1417190 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001384025.1 |
|
1 - 2 |
177 |
63 |
View Full Product Details