Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-64
Entrez Gene ID
3565142
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_Y22D7AR.8
Chromosome Location
Chr.III: 1692081 - 1695106 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065044.4 NP_497445.2 4 - 5 790 82

View Full Product Details