Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pvf-1
Entrez Gene ID
175327
Gene Name
PDGF_2 domain-containing protein
Gene Aliases
CELE_Y39A3CL.6
Chromosome Location
Chr.III: 1821149 - 1824314 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065060.4 NP_497461.1 4 - 5 759 76

View Full Product Details