Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-33
Entrez Gene ID
189728
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_Y39A3CR.6
Chromosome Location
Chr.III: 1855392 - 1859615 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368616.2 NP_001355375.1 1 - 2 255 118

View Full Product Details