Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
smi-1
Entrez Gene ID
189726
Gene Name
protein-coding
Gene Aliases
CELE_Y39A3CR.1
Chromosome Location
Chr.III: 1878365 - 1880866 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027676.4 NP_001022847.1 3 - 4 505 73
NM_001373850.1 NP_001359807.1 3 - 4 503 73

View Full Product Details