Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
feh-1
Entrez Gene ID
175373
Gene Name
protein-coding
Gene Aliases
CELE_Y54F10AM.2
Chromosome Location
Chr.III: 2549927 - 2560689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027735.5 NP_001022906.1 9 - 10 1708 89
NM_001373858.2 NP_001360128.1 7 - 8 1549 89
NM_001384041.1 8 - 9 1669 89
NM_065177.6 NP_497578.4 9 - 10 1713 89

View Full Product Details