Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-6
Entrez Gene ID
175462
Gene Name
protein-coding
Gene Aliases
CELE_C48D5.1
Chromosome Location
Chr.III: 3563684 - 3578770 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136364.4 NP_001129836.1 12 - 13 1837 71
NM_065330.7 NP_497731.1 12 - 13 1766 71
GenBank mRNA AF083224.1 - 12 - 13 1788 71
AY204167.1 - 11 - 12 1767 71

View Full Product Details