Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-4
Entrez Gene ID
175488
Gene Name
Galectin
Gene Aliases
CELE_C44F1.3
Chromosome Location
Chr.III: 3771425 - 3773394 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065362.8 NP_497763.1 3 - 4 369 124

View Full Product Details