Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-25A2
Entrez Gene ID
175495
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_C36A4.2
Chromosome Location
Chr.III: 3835221 - 3837211 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065375.3 NP_497776.2 2 - 3 237 141

View Full Product Details