Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbb-2
Entrez Gene ID
175519
Gene Name
Tubulin beta-2 chain
Gene Aliases
CELE_C36E8.5
Chromosome Location
Chr.III: 4015910 - 4017627 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065405.8 NP_497806.1 2 - 3 685 71

View Full Product Details