Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mcm-5
Entrez Gene ID
175552
Gene Name
DNA replication licensing factor mcm-5
Gene Aliases
CELE_R10E4.4
Chromosome Location
Chr.III: 4288262 - 4291266 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065457.6 NP_497858.1 2 - 3 495 77

View Full Product Details