Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clp-2
Entrez Gene ID
175618
Gene Name
Calpain catalytic domain-containing protein
Gene Aliases
CELE_T04A8.16
Chromosome Location
Chr.III: 4700325 - 4704054 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065563.4 NP_497964.1 5 - 6 988 146

View Full Product Details