Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-4
Entrez Gene ID
175643
Gene Name
protein-coding
Gene Aliases
CELE_C35D10.9
Chromosome Location
Chr.III: 4852325 - 4856203 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026031.4 NP_001021202.1 6 - 7 1311 65
NM_001026032.3 NP_001021203.1 6 - 7 1377 65

View Full Product Details