Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhl-2
Entrez Gene ID
175654
Gene Name
NHL (ring finger b-box coiled coil) domain containing
Gene Aliases
CELE_F26F4.7
Chromosome Location
Chr.III: 4893658 - 4899085 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065625.5 NP_498026.1 2 - 3 597 82

View Full Product Details