Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
klp-6
Entrez Gene ID
175683
Gene Name
Kinesin-like protein
Gene Aliases
CELE_R144.1
Chromosome Location
Chr.III: 5030929 - 5036583 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065668.6 NP_498069.1 4 - 5 1011 125
GenBank mRNA AB023497.1 - 4 - 5 1011 125

View Full Product Details