Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prmt-5
Entrez Gene ID
175717
Gene Name
Protein arginine N-methyltransferase 5
Gene Aliases
CELE_C34E10.5
Chromosome Location
Chr.III: 5230570 - 5234530 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065711.8 NP_498112.1 7 - 8 1921 128

View Full Product Details