Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gop-3
Entrez Gene ID
175723
Gene Name
SAM50-like protein gop-3
Gene Aliases
CELE_C34E10.1
Chromosome Location
Chr.III: 5260694 - 5263837 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065718.8 NP_498119.1 2 - 3 443 77
GenBank mRNA AY052771.1 - 2 - 3 443 77

View Full Product Details