Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ztf-8
Entrez Gene ID
175726
Gene Name
Zinc finger putative Transcription Factor family
Gene Aliases
CELE_ZC395.8
Chromosome Location
Chr.III: 5268579 - 5273471 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065723.6 NP_498124.2 7 - 8 1202 74

View Full Product Details