Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-23
Entrez Gene ID
189610
Gene Name
protein-coding
Gene Aliases
CELE_Y37B11A.1
Chromosome Location
Chr.III: 5770185 - 5774703 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330885.2 NP_001317785.1 2 - 3 208 134

View Full Product Details