Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpf-4
Entrez Gene ID
175821
Gene Name
Peptidase_S9 domain-containing protein
Gene Aliases
CELE_F01F1.5
Chromosome Location
Chr.III: 5859306 - 5861908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382883.1 4 - 5 1592 119

View Full Product Details