Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frm-2
Entrez Gene ID
175848
Gene Name
protein-coding
Gene Aliases
CELE_T04C9.6
Chromosome Location
Chr.III: 5976781 - 5982128 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047405.6 NP_001040870.1 9 - 10 1677 67
NM_001379736.1 NP_001367783.1 10 - 11 1772 67

View Full Product Details