Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
set-1
Entrez Gene ID
175918
Gene Name
protein-coding
Gene Aliases
CELE_T26A5.7
Chromosome Location
Chr.III: 6450792 - 6452378 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027625.4 NP_001022796.1 1 - 2 182 96
NM_001379764.2 NP_001367441.1 1 - 2 188 96

View Full Product Details