Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-10
Entrez Gene ID
182449
Gene Name
Prion-like-(Q/N-rich)-domain-bearing protein
Gene Aliases
CELE_C09E7.2
Chromosome Location
Chr.III: 6528202 - 6530517 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379768.1 NP_001367476.1 3 - 4 441 84

View Full Product Details