Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-40
Entrez Gene ID
191859
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_Y40D12A.3
Chromosome Location
Chr.III: 6610822 - 6612561 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066058.2 NP_498459.1 3 - 4 963 86

View Full Product Details