Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-41
Entrez Gene ID
175946
Gene Name
BTB and MATH domain-containing protein 41
Gene Aliases
CELE_F37A4.9
Chromosome Location
Chr.III: 6720413 - 6722258 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066072.7 NP_498473.1 4 - 5 466 94

View Full Product Details