Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rnp-7
Entrez Gene ID
176002
Gene Name
RRM domain-containing protein
Gene Aliases
CELE_K04G7.10
Chromosome Location
Chr.III: 7158912 - 7161155 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066164.8 NP_498565.2 2 - 3 334 79

View Full Product Details