Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-14
Entrez Gene ID
185423
Gene Name
Aldo_ket_red domain-containing protein
Gene Aliases
CELE_F37C12.12
Chromosome Location
Chr.III: 7190189 - 7192992 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066179.6 NP_498580.2 3 - 4 346 70

View Full Product Details