Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-13
Entrez Gene ID
176069
Gene Name
Homeobox protein ceh-13
Gene Aliases
CELE_R13A5.5
Chromosome Location
Chr.III: 7555070 - 7558208 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066254.7 NP_498655.1 3 - 4 534 119

View Full Product Details