Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mab-5
Entrez Gene ID
176091
Gene Name
Homeobox protein mab-5
Gene Aliases
CELE_C08C3.3
Chromosome Location
Chr.III: 7776835 - 7783461 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066294.7 NP_498695.1 1 - 2 217 84
GenBank mRNA AF277990.1 - 1 - 2 232 84

View Full Product Details