Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
prp-8
Entrez Gene ID
176153
Gene Name
Pre-mRNA-splicing factor 8 homolog
Gene Aliases
CELE_C50C3.6
Chromosome Location
Chr.III: 8164373 - 8173154 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066384.6 NP_498785.1 2 - 3 1074 68

View Full Product Details