Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-36
Entrez Gene ID
176155
Gene Name
protein-coding
Gene Aliases
CELE_C50C3.9
Chromosome Location
Chr.III: 8193837 - 8201217 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047386.5 NP_001040851.1 14 - 15 3529 102
NM_001268026.4 NP_001254955.1 13 - 14 3408 102

View Full Product Details