Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-2
Entrez Gene ID
259752
Gene Name
TetraSPanin family
Gene Aliases
CELE_C02F5.11
Chromosome Location
Chr.III: 8236928 - 8237935 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066402.2 NP_498803.1 1 - 2 124 101

View Full Product Details