Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cids-1
Entrez Gene ID
176162
Gene Name
CID domain-containing protein 1
Gene Aliases
CELE_C02F5.4
Chromosome Location
Chr.III: 8244289 - 8245800 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066406.6 NP_498807.2 5 - 6 763 83

View Full Product Details