Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sas-4
Entrez Gene ID
176172
Gene Name
Spindle assembly abnormal protein 4
Gene Aliases
CELE_F10E9.8
Chromosome Location
Chr.III: 8314345 - 8318588 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066427.7 NP_498828.2 9 - 10 2152 87
GenBank mRNA AJ539470.1 - 9 - 10 2145 87

View Full Product Details