Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutc-1
Entrez Gene ID
176186
Gene Name
Copper homeostasis protein cutC homolog
Gene Aliases
CELE_ZK353.7
Chromosome Location
Chr.III: 8402049 - 8403607 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066454.6 NP_498855.1 1 - 2 314 92

View Full Product Details