Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- pdhk-2
- Entrez Gene ID
-
176226
- Gene Name
- Probable [pyruvate dehydrogenase (acetyl-transferring)] kinase, mitochondrial
- Gene Aliases
-
CELE_ZK370.5
- Chromosome Location
-
Chr.III: 8737686 - 8741412 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_066527.6 |
NP_498928.1 |
1 - 2 |
60 |
97 |
View Full Product Details