Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tpk-1
Entrez Gene ID
176258
Gene Name
Thiamin pyrophosphokinase 1
Gene Aliases
CELE_ZK637.9
Chromosome Location
Chr.III: 8912144 - 8913632 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027853.2 NP_001023024.1 2 - 3 258 134

View Full Product Details