Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tax-4
Entrez Gene ID
176297
Gene Name
Cyclic nucleotide-gated cation channel
Gene Aliases
CELE_ZC84.2
Chromosome Location
Chr.III: 9181546 - 9185888 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066632.6 NP_499033.1 1 - 2 140 66
GenBank mRNA D86922.1 - 1 - 2 158 66

View Full Product Details