Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tkr-1
Entrez Gene ID
192055
Gene Name
Probable G-protein coupled receptor tkr-1
Gene Aliases
CELE_C38C10.1
Chromosome Location
Chr.III: 9383796 - 9387709 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066663.5 NP_499064.2 4 - 5 620 116

View Full Product Details